Skip to Content
Merck
All Photos(1)

Key Documents

797820

Sigma-Aldrich

2-{3-[ω-Methylpoly(dimethylsiloxane)-α-yl]propoxy}ethyl {2-[(2-methylprop-2-enoyl)oxy]ethyl}carbamate

contains ≤500 ppm phenothiazine as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H20NO5[C2H6OSi]nC2H6Si-R
UNSPSC Code:
12162002

form

liquid

contains

≤500 ppm phenothiazine as inhibitor

density

0.973 at 25 °C

storage temp.

2-8°C

Application

UV curable PDMS based monomer for silicone hydrogel application in medical devices.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service