Skip to Content
Merck
All Photos(1)

Key Documents

794457

Sigma-Aldrich

1,2:3,4-Di-O-isopropylidene-6-O-methacryloyl-α-D-galactopyranose

Synonym(s):

MAGP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O7
CAS Number:
Molecular Weight:
328.36
UNSPSC Code:
12162002
PubChem Substance ID:

form

solid

storage temp.

2-8°C

SMILES string

CC(C(OC[C@@H](O1)[C@H](O2)[C@H](OC2(C)C)[C@@H](O3)[C@H]1OC3(C)C)=O)=C

InChI

1S/C16H24O7/c1-8(2)13(17)18-7-9-10-11(21-15(3,4)20-10)12-14(19-9)23-16(5,6)22-12/h9-12,14H,1,7H2,2-6H3/t9-,10+,11+,12-,14-/m1/s1

InChI key

RKFOJTMVZFAYQE-OIRVYTLQSA-N

Application

This monomer can be used to make macromolecules containing carbohydrate moieties. Controlled Radical Polymerization; such as NMP; RAFT; or ATRP; is commonly employed to synthesize well-defined glycopolymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ladmiral; Vincent;
Polymer, 46(19), 8536-8545 (2005)
Carneiro; M. J.;
Carbohydrate Polymers, 45(2), 135-138 (2001)
Lowe; Andrew B.;
Polymer, 48(8), 2221-2230 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service