Skip to Content
Merck
All Photos(1)

Key Documents

793744

Sigma-Aldrich

Benzofuran-d6

97 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D6O
Molecular Weight:
124.17
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

97% (CP)

form

liquid

mass shift

M+6

storage temp.

2-8°C

SMILES string

[2H]C1=C([2H])C([2H])=C([2H])C2=C1C([2H])=C([2H])O2

InChI

1S/C8H6O/c1-2-4-8-7(3-1)5-6-9-8/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

IANQTJSKSUMEQM-MZWXYZOWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service