All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H6F2O2S
CAS Number:
Molecular Weight:
192.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
mp
25-30 °C
SMILES string
CC1=C(S(F)(=O)=O)C=C(F)C=C1
InChI
1S/C7H6F2O2S/c1-5-2-3-6(8)4-7(5)12(9,10)11/h2-4H,1H3
InChI key
YWDLFHAWBWCNTB-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service