Skip to Content
Merck
All Photos(1)

Key Documents

78803

Sigma-Aldrich

(+)-8-Phenylmenthol

≥98.0% (sum of enantiomers, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O
CAS Number:
Molecular Weight:
232.36
Beilstein:
5265529
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (sum of enantiomers, HPLC)

form

liquid

optical activity

[α]20/D +26±1°, c = 2% in ethanol

refractive index

n20/D 1.531

density

0.999 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H]1CC[C@@H]([C@@H](O)C1)C(C)(C)c2ccccc2

InChI

1S/C16H24O/c1-12-9-10-14(15(17)11-12)16(2,3)13-7-5-4-6-8-13/h4-8,12,14-15,17H,9-11H2,1-3H3/t12-,14-,15-/m0/s1

InChI key

WTQIZFCJMGWUGZ-QEJZJMRPSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service