Skip to Content
Merck
All Photos(1)

Key Documents

776920

Sigma-Aldrich

Thiophene-2,5-diboronic acid bis(pinacol) ester

99% (GC)

Synonym(s):

2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene, 2,5-Thiophenediboronic acid bis(pinacol) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26B2O4S
CAS Number:
Molecular Weight:
336.06
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99% (GC)

form

powder

mp

227-231 °C

InChI

1S/C16H26B2O4S/c1-13(2)14(3,4)20-17(19-13)11-9-10-12(23-11)18-21-15(5,6)16(7,8)22-18/h9-10H,1-8H3

InChI key

AOJXAKMKFDBHHT-UHFFFAOYSA-N

Application

Diboronic acid ester thiophene monomer for polymerization by Suzuki coupling; used for OPV and OFET applications.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee; Joong Suk;
Chemistry of Materials, 24, 1316-1323 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service