Skip to Content
Merck
All Photos(1)

Documents

769401

Sigma-Aldrich

Hafnium(IV) oxide

pellets, diam. × thickness 13 mm × 5 mm

Synonym(s):

Hafnia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HfO2
CAS Number:
Molecular Weight:
210.49
EC Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

form

pellets

diam. × thickness

13 mm × 5 mm

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

density

9.68 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.0005%
Cr: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.005%
Hg: ≤0.1%
Pb: ≤0.1%
Ti: ≤0.005%
V: ≤0.005%
Zr: ≤0.5%

SMILES string

O=[Hf]=O

InChI

1S/Hf.2O

InChI key

CJNBYAVZURUTKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High dielectric constant of hafnia makes it suitable for application in metal oxide based semiconductor devices. It also finds applications as optical coating materials

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent progress in ab initio simulations of hafnia-based gate stacks
Zhu, H., et al.
J. Mater. Sci., 47(21), 7399-7416 (2012)
Dual wavelength laser-induced damage threshold measurements of alumina/silica and hafnia/silica ultraviolet antireflective coatings.
Mrohs, M., et al.
Applied Optics, 55(1), 104-109 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service