Skip to Content
Merck
All Photos(1)

Documents

766720

Sigma-Aldrich

2,4,7,8,9-Penta-O-acetyl-N-acetylneuraminic acid methyl ester

97%

Synonym(s):

N-Acetylneuraminic acid methyl ester 2,4,7,8,9-pentaacetate, Methyl 2,4,7,8,9-penta-O-acetyl-N-acetylneuraminate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO14
CAS Number:
Molecular Weight:
533.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

form

solid

mp

154-155 °C (lit.)
154-155 °C

storage temp.

−20°C

SMILES string

CC(O[C@]1(C(OC)=O)O[C@]([C@H](OC(C)=O)[C@H](OC(C)=O)COC(C)=O)([H])[C@H](NC(C)=O)[C@@H](OC(C)=O)C1)=O

InChI

1S/C22H31NO14/c1-10(24)23-18-16(33-12(3)26)8-22(21(30)31-7,36-15(6)29)37-20(18)19(35-14(5)28)17(34-13(4)27)9-32-11(2)25/h16-20H,8-9H2,1-7H3,(H,23,24)/t16-,17+,18+,19+,20+,22-/m0/s1

InChI key

MFDZYSKLMAXHOV-QDRUEPRISA-N

Application

Versatile acetylated neuraminic acid or sialic acid derivative. N-acetylneuraminic acid is often present as the terminal sugar of glycoproteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service