Skip to Content
Merck
All Photos(1)

Documents

755834

Sigma-Aldrich

2,2′-Diallyl bisphenol A diacetate ether

90%

Synonym(s):

EC-392, Phenyl ester epoxy curative hybrid of bisphenol A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28O4
CAS Number:
Molecular Weight:
392.49
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

form

liquid

refractive index

n20/D 1.548

viscosity

~3500 cP(25 °C) (Haake C/P Rheometer, 5 RPM)

density

1.096 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc(cc1CC=C)C(C)(C)c2ccc(OC(C)=O)c(CC=C)c2

InChI

1S/C25H28O4/c1-7-9-19-15-21(11-13-23(19)28-17(3)26)25(5,6)22-12-14-24(29-18(4)27)20(16-22)10-8-2/h7-8,11-16H,1-2,9-10H2,3-6H3

InChI key

GJTZQXNRNNDGRG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service