Skip to Content
Merck
All Photos(1)

Key Documents

754528

Sigma-Aldrich

9-[2-(Dicyclohexylphosphino)phenyl]-9H-carbazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H34NP
CAS Number:
Molecular Weight:
439.57
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

form

solid

reaction suitability

reagent type: ligand

mp

186-190 °C

functional group

phosphine

SMILES string

C1CCC(CC1)P(C2CCCCC2)c3ccccc3-n4c5ccccc5c6ccccc46

InChI

1S/C30H34NP/c1-3-13-23(14-4-1)32(24-15-5-2-6-16-24)30-22-12-11-21-29(30)31-27-19-9-7-17-25(27)26-18-8-10-20-28(26)31/h7-12,17-24H,1-6,13-16H2

InChI key

DGWSERVYJXYEDY-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service