Skip to Content
Merck
All Photos(1)

Key Documents

752541

Sigma-Aldrich

2,5-Dibromo-3,4-dihexylthiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26Br2S
CAS Number:
Molecular Weight:
410.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.534

density

1.312 g/mL at 25 °C

SMILES string

CCCCCCc1c(Br)sc(Br)c1CCCCCC

InChI

1S/C16H26Br2S/c1-3-5-7-9-11-13-14(12-10-8-6-4-2)16(18)19-15(13)17/h3-12H2,1-2H3

InChI key

YJMJDAXLNHAHAQ-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service