Skip to Content
Merck
All Photos(1)

Key Documents

741426

Sigma-Aldrich

Tributyl (1,3-dioxolan-2-ylmethyl)phosphonium bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H34BrO2P
Molecular Weight:
369.32
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

25-40 °C

functional group

ether
phosphine

storage temp.

2-8°C

SMILES string

[Br-].CCCC[P+](CCCC)(CCCC)CC1OCCO1

InChI

1S/C16H34O2P.BrH/c1-4-7-12-19(13-8-5-2,14-9-6-3)15-16-17-10-11-18-16;/h16H,4-15H2,1-3H3;1H/q+1;/p-1

InChI key

SABIECHYMHKXJI-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service