Skip to Content
Merck
All Photos(1)

Key Documents

731293

Sigma-Aldrich

N-Boc-hexamethyleneimine

97%

Synonym(s):

1-(tert-Butoxycarbonyl)hexahydroazepine, 1-Boc-azepane, 1-Boc-hexahydro-1H-azepine, 1-Boc-homopiperidine, 1-Boc-perhydroazepine, N-Boc-azepane, N-Boc-perhydroazepine, Hexahydro-1H-azepine-1-carboxylic acid 1,1-dimethylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO2
CAS Number:
Molecular Weight:
199.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.457

density

0.967 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N1CCCCCC1

InChI

1S/C11H21NO2/c1-11(2,3)14-10(13)12-8-6-4-5-7-9-12/h4-9H2,1-3H3

InChI key

QZBSPLNCSRGMBA-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

221.0 °F

Flash Point(C)

105 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service