Skip to Content
Merck
All Photos(1)

Key Documents

729655

Sigma-Aldrich

Acetylene-13C2 dicarboxylic acid

99 atom % 13C, 98% (CP)

Synonym(s):

2-Butynedioic acid-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C13C13CCO2H
CAS Number:
Molecular Weight:
116.04
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+2

storage temp.

2-8°C

SMILES string

OC(=O)[13C]#[13C]C(O)=O

InChI

1S/C4H2O4/c5-3(6)1-2-4(7)8/h(H,5,6)(H,7,8)/i1+1,2+1

InChI key

YTIVTFGABIZHHX-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service