Skip to Content
Merck
All Photos(1)

Key Documents

725048

Sigma-Aldrich

3-Aminopyridazine

97%

Synonym(s):

Pyridazin-3-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3
CAS Number:
Molecular Weight:
95.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

168-172 °C

SMILES string

Nc1cccnn1

InChI

1S/C4H5N3/c5-4-2-1-3-6-7-4/h1-3H,(H2,5,7)

InChI key

LETVJWLLIMJADE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Somayeh Pirhadi et al.
Journal of receptor and signal transduction research, 40(4), 357-364 (2020-03-05)
Everyday plenty of people succumb to various forms of cancer across the world and it stands as one of the main reasons of death in our today's life. Receptor tyrosine kinases (RTKs) are a class of receptors involved in cancer
Shawna D Persaud et al.
Scientific reports, 6, 22396-22396 (2016-03-05)
All trans retinoic acid (atRA) is one of the most potent therapeutic agents, but extensive toxicity caused by nuclear RA receptors (RARs) limits its clinical application in treating cancer. AtRA also exerts non-genomic activities for which the mechanism remains poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service