Skip to Content
Merck
All Photos(1)

Key Documents

709352

Sigma-Aldrich

Cyclopropyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NS
CAS Number:
Molecular Weight:
99.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.552

density

1.053 g/mL at 25 °C

functional group

isothiocyanate

storage temp.

2-8°C

SMILES string

S=C=NC1CC1

InChI

1S/C4H5NS/c6-3-5-4-1-2-4/h4H,1-2H2

InChI key

JGFBQFKZKSSODQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.9 °F - closed cup

Flash Point(C)

54.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service