Skip to Content
Merck
All Photos(1)

Key Documents

704687

Sigma-Aldrich

5-Pyrimidineboronic acid pinacol ester

97%

Synonym(s):

5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15BN2O2
CAS Number:
Molecular Weight:
206.05
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

mp

67-71 °C

SMILES string

CC1(C)OB(OC1(C)C)c2cncnc2

InChI

1S/C10H15BN2O2/c1-9(2)10(3,4)15-11(14-9)8-5-12-7-13-6-8/h5-7H,1-4H3

InChI key

WSRGGSAGSUEJKQ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service