Skip to Content
Merck
All Photos(1)

Documents

701467

Sigma-Aldrich

1-(4-Morpholinyl)-1-propanone

97%

Synonym(s):

1-Morpholino-1-propanone, 4-(1-Oxopropyl)morpholine, 4-Propionylmorpholine, N-Propanoylmorpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

bp

70-75 °C/0.5 mmHg

density

1.068 g/mL at 25 °C

SMILES string

CCC(=O)N1CCOCC1

InChI

1S/C7H13NO2/c1-2-7(9)8-3-5-10-6-4-8/h2-6H2,1H3

InChI key

DLEWDCPFCNLJEY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel A Lago et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 33(3), 518-529 (2016-01-21)
Since the UV ink photoinitiator (PI) isopropylthioxanthone (ITX) was discovered in packaged milk, studies of print contamination have focused primarily on PIs but have also included amine synergists. Many other substances are used or formed during the print process, yet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service