669784
Methyl 6-hydroxy-5-nitropyridine-3-carboxylate
97%
Synonym(s):
6-Hydroxy-5-nitronicotinic acid methyl ester, Methyl 6-hydroxy-5-nitronicotinate
About This Item
Recommended Products
Assay
≥96.5% (HPLC)
97%
functional group
ester
nitro
SMILES string
COC(=O)c1cnc(O)c(c1)[N+]([O-])=O
InChI
1S/C7H6N2O5/c1-14-7(11)4-2-5(9(12)13)6(10)8-3-4/h2-3H,1H3,(H,8,10)
InChI key
ILGFGEIQCLRIBI-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service