Skip to Content
Merck
All Photos(1)

Documents

648299

Sigma-Aldrich

PAMAM-tris(hydroxymethyl)amidomethane dendrimer, 1,12-diaminododecane core, generation 6 solution

10 wt. % in H2O

Synonym(s):

PAMAM-tris(hydroxymethyl)amidomethane dendrimer, 1,12-diaminododecane core, generation 6 solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)12NH2]:(G=6);dendri PAMAM[NHC(CH2OH)3]256
Molecular Weight:
73811.78
MDL number:
UNSPSC Code:
12162002

description

tris(hydroxymethyl)amidomethane surface groups

form

solution

mol wt

generation 6

feature

core type 1,12-diaminododecane core (12-carbon core)

concentration

10 wt. % in H2O

no. Surface Groups

256

bp

100 °C

density

1.00 g/mL at 25 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Contains 256 surface tris(hydroxymethyl)aminomethane groups.

Packaging

unit weight includes solvent

Legal Information

Mfg. by Dendritic NanoTechnologies Inc. Licensed for research use only.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service