64297
2,2,2-Trifluoroethyl methanesulfonate
≥98.0% (GC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3SO3CH2CF3
CAS Number:
Molecular Weight:
178.13
Beilstein:
1863049
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.0% (GC)
refractive index
n20/D 1.367
density
1.513 g/mL at 20 °C (lit.)
SMILES string
CS(=O)(=O)OCC(F)(F)F
InChI
1S/C3H5F3O3S/c1-10(7,8)9-2-3(4,5)6/h2H2,1H3
InChI key
ICECLJDLAVVEOW-UHFFFAOYSA-N
Other Notes
Reagent for 2,2,2-trifluoroethylations, e.g. ether formation
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
219.2 °F - closed cup
Flash Point(C)
104 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
F. Camps et al.
Synthesis, 727-727 (1980)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service