Skip to Content
Merck
All Photos(1)

Documents

640220

Sigma-Aldrich

Poly(vinyl phenyl sulfide-co-glycidyl methacrylate)

Glycidyl methacrylate 10 mol %

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

composition

Glycidyl methacrylate, 10 mol %

refractive index

n20/D 1.6568 (lit.)

λmax

221 nm

General description

High refractive index polymer

Application

Photocrosslinkable polymer for optical waveguides

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Refractive Index Polymers for Optical Applications
Beecroft, L.L and Ober, C.K.
J. Macromol. Sci., Pure Appl. Chem., A34, 573-586 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service