Skip to Content

2025 - 1月及農曆春節預計貨物安排,請參考此頁訊息

Merck
All Photos(1)

Key Documents

637165

Sigma-Aldrich

Diethyl-3,4-ethylenedioxypyrrole-2,5-dicarboxylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO6
CAS Number:
Molecular Weight:
269.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

98%

mp

124-128 °C (lit.)

SMILES string

CCOC(=O)c1[nH]c(C(=O)OCC)c2OCCOc12

InChI

1S/C12H15NO6/c1-3-16-11(14)7-9-10(19-6-5-18-9)8(13-7)12(15)17-4-2/h13H,3-6H2,1-2H3

InChI key

PRLDMMBRDYVWRG-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF10253179GF96137837GF78358438
铜 foil, 25mm disks, thickness 0.075mm, half hard, 99.95+%

GF03625051

铜 foil, 25mm disks, thickness 0.7mm, half hard, 99.95+%

GF10253179

铜 foil, 25mm disks, thickness 1.0mm, half hard, 99.95+%

GF96137837

铜 foil, 25mm disks, thickness 0.25mm, half hard, 99.95+%

GF78358438

assay

99.95%

assay

≥99.95%

assay

≥99.95%

assay

99.95%

form

foil

form

foil

form

foil

form

foil

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

Application

Starting monomer for a variety of etheylenedioxypyrrole (EDOP) conductive and electrooptical polymers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schottland, P.; Reynolds, J.;
Macromolecules, 33, 7051-7051 (2000)
K Zong et al.
The Journal of organic chemistry, 66(21), 6873-6882 (2001-10-13)
New functionalized derivatives of 3,4-ethylenedioxypyrrole (EDOP, 5a) and 3,4-(1,3-propylenedioxy)pyrrole (ProDOP, 5b) as especially electron-rich monomers which yield highly electroactive and stable conducting polymers useful for a diverse set of applications have been synthesized. N-Alkylations of ProDOP were carried out to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service