Skip to Content
Merck
All Photos(1)

Documents

609951

Sigma-Aldrich

Silicon dioxide-18O2

95 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si18O2
Molecular Weight:
64.08
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:

isotopic purity

95 atom % 18O

form

solid

mass shift

M+4

SMILES string

[18O]=[Si]=[18O]

InChI

1S/O2Si/c1-3-2/i1+2,2+2

InChI key

VYPSYNLAJGMNEJ-XPULMUKRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service