Skip to Content
Merck
All Photos(1)

Key Documents

562016

Sigma-Aldrich

4-Fluorobenzylmagnesium chloride solution

0.25 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2MgCl
CAS Number:
Molecular Weight:
168.88
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.25 M in diethyl ether

bp

35 °C

density

0.742 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Fc1ccc(C[Mg]Cl)cc1

InChI

1S/C7H6F.ClH.Mg/c1-6-2-4-7(8)5-3-6;;/h2-5H,1H2;1H;/q;;+1/p-1

InChI key

PQDGQUPDDGUKLP-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service