Skip to Content
Merck
All Photos(1)

Key Documents

561762

Sigma-Aldrich

4-Thioanisolemagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CSC6H4MgBr
CAS Number:
Molecular Weight:
227.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

65 °C

density

0.965 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CSc1ccc([Mg]Br)cc1

InChI

1S/C7H7S.BrH.Mg/c1-8-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

SSVQCUSLJVVRCA-UHFFFAOYSA-M

Application

4-Thioanisolemagnesium bromide can be used to prepare:
  • 3,3-Ethylenedioxy-5a-hydroxy-11b-[4-(thiomethoxy)phenyl]-estr-9-en-17-one, a key intermediate for the synthesis of mifepristone analogs.[1]
  • (1S)-1,4-Anhydro-2,3,5,6-tetra-O-benzyl-1-C-[4-chloro-3-(4-methylthiobenzyl)-phenyl]-D-glucitol, a key intermediate for the synthesis of aryl D-glucofuranosides as potent hSGLT2 and hSGLT1 inhibitors.[2]

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzung-Sheng Lin et al.
Bioorganic & medicinal chemistry, 21(21), 6282-6291 (2013-09-28)
Novel C-aryl-d-glucofuranosides were synthesized and evaluated for their capacity to inhibit human sodium-dependent glucose co-transporter 2 (hSGLT2) and hSGLT1. Compound 21q demonstrated the best in vitro inhibitory activity against SGLT2 in this series (EC50=0.62μM).
Weiqin Jiang et al.
Steroids, 71(11-12), 949-954 (2006-08-29)
A novel series of steroidal compounds were designed and synthesized with various phosphorus-containing groups on the 17beta-side chain as progesterone receptor antagonists. The structure-activity relationships of these compounds are discussed. Selected compounds were tested in an rat progesterone-sensitive assay. Some

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service