Skip to Content
Merck
All Photos(1)

Documents

561665

Sigma-Aldrich

3-Fluorophenylmagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4MgBr
CAS Number:
Molecular Weight:
199.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

65 °C

density

0.936 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Fc1cccc([Mg]Br)c1

InChI

1S/C6H4F.BrH.Mg/c7-6-4-2-1-3-5-6;;/h1-2,4-5H;1H;/q;;+1/p-1

InChI key

VFPDAAQAKDGSHQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

3-Fluorophenylmagnesium bromide is a Grignard reagent that can be used in:
  • The preparation of 4-substituted quinazolines by reacting with 2-methylaminobenzonitriles.
  • The synthesis of radiolabeled TrkB/C-targeting kinase inhibitor for positron emission tomography (PET) imaging applications.
  • Cu-catalyzed carbometalation of substituted cyclopropenes.

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facially selective and regioselective carbometalation of cyclopropenes by aryl Grignard reagents
Yan N, et al.
The Journal of Organic Chemistry, 73(2), 563-568 (2008)
A kinome-wide selective radiolabeled TrkB/C inhibitor for in vitro and in vivo neuroimaging: synthesis, preclinical evaluation, and first-in-human
Bernard-Gauthier V, et al.
Journal of medicinal chemistry, 60(16), 6897-6910 (2017)
Synthesis of quinazolines via an iron-catalyzed oxidative amination of N-H ketimines
Chen C, et al.
The Journal of Organic Chemistry, 83(4), 2395-2401 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service