561282
4-Methoxybenzyl bromide
Synonym(s):
1-(Bromomethyl)-4-methoxybenzene, 4-(Bromomethyl)anisole, p-(Bromomethyl)anisole, p-Methoxybenzyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH2BrC6H4OCH3
CAS Number:
Molecular Weight:
201.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
contains
~3 wt. % K2CO3 as stabilizer
refractive index
n20/D 1.5780 (lit.)
bp
91 °C/1 mmHg (lit.)
density
1.395 g/mL at 19 °C (lit.)
SMILES string
COc1ccc(CBr)cc1
InChI
1S/C8H9BrO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6H2,1H3
InChI key
GIGRWGTZFONRKA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reagent for protection of hydroxyl groups that is removable by treatment with DDQ. Used in a recent synthesis of (R)-(–)-argentilactone.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis, 2879-2879 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service