Skip to Content
Merck
All Photos(2)

Documents

557781

Sigma-Aldrich

N-Benzyl-2,3-dibromomaleimide

97%

Synonym(s):

1-Benzyl-3,4-dibromo-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7Br2NO2
CAS Number:
Molecular Weight:
344.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

117-120 °C (lit.)

SMILES string

BrC1=C(Br)C(=O)N(Cc2ccccc2)C1=O

InChI

1S/C11H7Br2NO2/c12-8-9(13)11(16)14(10(8)15)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

DZYLZHRCCNTQCS-UHFFFAOYSA-N

Application

N-Benzyl-2,3-dibromomaleimide may be used in the synthesis of the following maleimide-based dyes, which can be employed in fluorescence quenching:
  • 2,3-bis(3-indolyl)-N-benzylmaleimide
  • 2,3-bis(2′-methyl-3-indolyl)-N-benzylmaleimide
  • 2,3-bis(2-pyrrolyl)-N-benzylmaleimide
  • 2-aryl-N-benzyl-3-bromomaleimides via suzuki cross coupling reaction with aryl boronic acid

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bifunctional maleimide dyes as selective anion sensors.
Lin Z, et al.
Tetrahedron, 65(27), 5216-5221 (2009)
"Metal-catalyzed cross-coupling reactions of halomaleic anhydrides and halomaleimides: synthesis of structurally interesting and biologically important natural and unnatural products"
Deore SP and Argade PN
Synthesis, 46(03), 281-289 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service