Skip to Content
Merck
All Photos(1)

Documents

557471

Sigma-Aldrich

4-Nitrothioanisole

96%

Synonym(s):

Methyl-4-nitrophenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SCH3
CAS Number:
Molecular Weight:
169.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

66-69 °C (lit.)
68-72 °C (lit.)

SMILES string

CSc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H7NO2S/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3

InChI key

NEZGPRYOJVPJKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrothioanisole in acetone-d6 solution exhibits twofold barrier to rotation about the Csp2-S bond as 16.1±1.5kJ/mol. 4-Nitrothioanisole undergoes hydrogenation in the presence of sulfided Pd/C catalysts.

Application

4-Nitrothioanisole may be used to synthesize 4-nitrothioanisole sulfoxide and methyl 4-nitrophenyl sulfoxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Horseradish peroxidase-catalyzed two-electron oxidations. Oxidation of iodide, thioanisoles, and phenols at distinct sites.
Harris RZ, et al.
The Journal of Biological Chemistry, 268(3), 1637-1645 (1993)
Catalytic hydrogenation of sulfur-containing nitrobenzene over Pd/C catalysts: In situ sulfidation of Pd/C for the preparation of Pd x S y catalysts.
Zhang Q, et al.
Applied Catalysis A: General, 497, 17-21 (2015)
Photoelectrochemical reduction of meta-halonitrobenzenes and related species.
Robert AW and George WJ.
J. Chem. Soc. Perkin Trans. II, 8, 1673-1677 (1995)
The proximate coupling constant, 5 J (H, CH3), and the torsional mobility of the thiomethyl group in some thioanisole derivatives.
Schaefer T, et al.
Canadian Journal of Chemistry, 69(4), 620-624 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service