Recommended Products
Assay
97%
mp
67-70 °C (lit.)
SMILES string
Fc1ccc(Cl)cc1C#N
InChI
1S/C7H3ClFN/c8-6-1-2-7(9)5(3-6)4-10/h1-3H
InChI key
GJNJDELEHIGPKJ-UHFFFAOYSA-N
General description
5-Chloro-2-fluorobenzonitrile is a benzonitrile derivative. The X-ray diffraction analysis indicates that its crystals belong to the monoclinic crystal system and P21/c space group.
5-Chloro-2-fluorobenzonitrile may be used in the preparation of methyl 3-amino-5-chlorobenzo[b]thiophene-2-carboxylate by reacting with methyl thioglycolate and triethylamine via microwave-assisted synthesis.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analysis of Small Molecule X-Ray Crystal Structures: Chemical Crystallography with Undergraduate Students in a Teaching Laboratory.
Journal of Chemical Crystallography, 44(2), 70-81 (2014)
Organic & biomolecular chemistry, 13(24), 6814-6824 (2015-05-28)
Microwave irradiation of 2-halobenzonitriles and methyl thioglycolate in the presence of triethylamine in DMSO at 130 °C provides rapid access to 3-aminobenzo[b]thiophenes in 58-96% yield. This transformation has been applied in the synthesis of the thieno[2,3-b]pyridine core motif of LIMK1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service