Skip to Content
Merck
All Photos(1)

Key Documents

539848

Sigma-Aldrich

5-(2-Furyl)-1,3-cyclohexanedione

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O3
CAS Number:
Molecular Weight:
178.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

152-155 °C (lit.)

SMILES string

O=C1CC(CC(=O)C1)c2ccco2

InChI

1S/C10H10O3/c11-8-4-7(5-9(12)6-8)10-2-1-3-13-10/h1-3,7H,4-6H2

InChI key

FYLTVHCMIYGVPZ-UHFFFAOYSA-N

General description

5-(2-Furyl)-1,3-cyclohexanedione may be used as a reactant in the synthesis of 12-aryl(hetaryl)-9-(2-furyl)-7,8,9,10,11,12-hexahydrobenzo[a]-acridin-11-ones bearing two asymmetric carbon atoms (C9 and C12).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three-component condensation of 2-naphthylamine with aromatic aldehydes and 5-(2-furyl)-1, 3-cyclohexanedione.
Kozlov, N. G., K. N. Gusak, and V. V. Skatetskii.
Russ. J. Org. Chem., 42(1), 112-117 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service