Skip to Content
Merck
All Photos(1)

Key Documents

537799

Sigma-Aldrich

1,2,3,4-Tetrahydrocyclopent[b] indole

96%

Synonym(s):

2,3-Trimethyleneindole, NSC 112674

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

100.5-105.5 °C (lit.)

SMILES string

C1Cc2[nH]c3ccccc3c2C1

InChI

1S/C11H11N/c1-2-6-10-8(4-1)9-5-3-7-11(9)12-10/h1-2,4,6,12H,3,5,7H2

InChI key

HZDXFZHFEASSBM-UHFFFAOYSA-N

General description

1,2,3,4-Tetrahydrocyclopent[b] indole undergoes reduction in the presence of Pd/C and hydrogen gas to yield 1,2,3,3a,4,8b-hexahydrocyclopent[b]indole.

Application

  • Reactant for thionium species-mediated functionalization at the 2α-position of indoles
  • Reactant for preparation of tetramethylpiperidine-1-oxoammonium salts as oxidants

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1, 2, 3, 4-tetrahydrocarbazoles and related tricyclic indoles.
Scott TL, et al.
Tetrahedron, 63(5), 1183-1183 (3007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service