Skip to Content
Merck
All Photos(1)

Key Documents

534161

Sigma-Aldrich

2-Amino-4-(p-tolyl)thiazole

96%

Synonym(s):

4-(4-Methylphenyl)-2-thiazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2S
CAS Number:
Molecular Weight:
190.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

132-136 °C (lit.)

SMILES string

Cc1ccc(cc1)-c2csc(N)n2

InChI

1S/C10H10N2S/c1-7-2-4-8(5-3-7)9-6-13-10(11)12-9/h2-6H,1H3,(H2,11,12)

InChI key

ARLHWYFAPHJCJT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service