concentration
0.5 M in THF
impurities
1-2% 4-(iodozinc)benzylzinc bromide
1-3% inorganic chloride
bp
65 °C
density
1.035 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
Br[Zn]Cc1ccc(I)cc1
InChI
1S/C7H6I.BrH.Zn/c1-6-2-4-7(8)5-3-6;;/h2-5H,1H2;1H;/q;;+1/p-1
InChI key
UKJFBRLWZXEWBX-UHFFFAOYSA-M
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
1.0 °F - closed cup
Flash Point(C)
-17.22 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service