Skip to Content
Merck
All Photos(1)

Key Documents

525103

Sigma-Aldrich

Tri(ethylene glycol) bis(2-ethylhexanoate)

97%

Synonym(s):

TEG(EH)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CO2CH2CH2OCH2-]2
CAS Number:
Molecular Weight:
402.57
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.445

bp

344 °C (lit.)

mp

−50 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)C(=O)OCCOCCOCCOC(=O)C(CC)CCCC

InChI

1S/C22H42O6/c1-5-9-11-19(7-3)21(23)27-17-15-25-13-14-26-16-18-28-22(24)20(8-4)12-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

FRQDZJMEHSJOPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service