Skip to Content
Merck
All Photos(1)

Key Documents

523100

Sigma-Aldrich

5-Fluoro-2-methoxybenzaldehyde

96%

Synonym(s):

5-Fluoro-o-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
154.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

mp

41-46 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

COc1ccc(F)cc1C=O

InChI

1S/C8H7FO2/c1-11-8-3-2-7(9)4-6(8)5-10/h2-5H,1H3

InChI key

CRLDWFVRQNUUSZ-UHFFFAOYSA-N

General description

5-Fluoro-2-methoxybenzaldehyde is an aryl fluorinated building block.

Application

5-Fluoro-2-methoxybenzaldehyde may be used in the synthesis of 5-(5-fluoro-2-methoxyphenyl)imidazolidine-2,4-dione.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Czopek et al.
Archiv der Pharmazie, 346(2), 98-109 (2013-01-05)
A computer aided ligand design study of imidazolidine-2,4-dione derivatives was conducted in order to obtain compounds with dual 5-HT(1A) receptor and serotonin transporter (SERT) affinity. According to molecular modeling results, series of Mannich bases were chosen and synthesized. Investigated compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service