Skip to Content
Merck
All Photos(1)

Documents

472298

Sigma-Aldrich

2,6-Dichloro-5-fluoropyridine-3-carboxylic acid

97%

Synonym(s):

2,6-Dichloro-5-fluoro-3-pyridinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl2FNO2
CAS Number:
Molecular Weight:
209.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

152-155 °C (lit.)

SMILES string

OC(=O)c1cc(F)c(Cl)nc1Cl

InChI

1S/C6H2Cl2FNO2/c7-4-2(6(11)12)1-3(9)5(8)10-4/h1H,(H,11,12)

InChI key

LTDGKGCHRNNCAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dichloro-5-fluoropyridine-3-carboxylic acid is a halogenated heterocyclic compound. Its density has been reported to be 1.6207g/cm3 at 25°C.

Application

2,6-Dichloro-5-fluoropyridine-3-carboxylic acid (2,6-Dichloro-5-fluoro-3-pyridinecarboxylic acid) may be used in the preparation of self assembled monolayer (SAMs) compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The adhesive properties of pyridine-terminated self-assembled monolayers.
Bowen J, et al.
Thin Solid Films, 517(13), 3806-3812 (2009)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 88-88 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service