Skip to Content
Merck
All Photos(1)

Key Documents

468991

Sigma-Aldrich

2-Nitrophenyl phenyl sulfide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SC6H5
CAS Number:
Molecular Weight:
231.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

78-82 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1Sc2ccccc2

InChI

1S/C12H9NO2S/c14-13(15)11-8-4-5-9-12(11)16-10-6-2-1-3-7-10/h1-9H

InChI key

ZPWNCSAEXUDWTN-UHFFFAOYSA-N

General description

2-Nitrophenyl phenyl sulfide is an aromatic sulfide bearing a nitro group. It has been reported to undergo sulfur oxidation on electrospray ionization in the positive-ion mode. Accurate masses of fragment ions generated by electrospray ionization (ESI) - collisionally activated dissociation (CAD) of [M + H]+ ions of 2-nitrophenyl phenyl sulfide have been evaluated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph T Moolayil et al.
Journal of the American Society for Mass Spectrometry, 18(12), 2204-2217 (2007-11-03)
A novel gas-phase electrophilic cyclization, initiated by the protonation of a nitro group, occurs for 2-nitrophenyl phenyl ether and for the analogous sulfide and amine, leading to heterocyclic intermediates in each case. Subsequently, the cyclic intermediates dissociate via two pathways:
Joseph T Moolayil et al.
International journal of mass spectrometry, 283(1-3), 222-228 (2010-02-18)
Aromatic sulfides bearing a nitro group undergo sulfur oxidation upon electrospray ionization in the positive-ion mode. For example, 2-nitrophenyl phenyl sulfide, its para nitro isomer, and its chloro and methyl substituted analogs pick up an oxygen atom to afford [M
Photochemical Rearrangement of 2-Nitrophenyl Phenyl Sulfoxide to 2-Nitrosophenyl Phenyl Sulfone.
Tanikaga R and Kaji A.
Bulletin of the Chemical Society of Japan, 46(12), 3814-3817 (1973)
Ellen K Parker et al.
Analytical and bioanalytical chemistry, 411(21), 5405-5413 (2018-11-02)
Preterm birth (PTB) is defined as birth before the 37th week of pregnancy and results in 15 million early deliveries worldwide every year. Presently, there is no clinical test to determine PTB risk; however, a panel of nine biomarkers found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service