463256
Glycolic acid ethoxylate lauryl ether
average Mn ~690
Synonym(s):
Laureth-11 carboxylic acid
About This Item
Recommended Products
mol wt
average Mn ~690
impurities
≤1.5% NaCl
8-12% water
refractive index
n20/D 1.454
density
1 g/mL at 25 °C (lit.)
SMILES string
OCCO.OCC(O)=O.CCCCCCCCCCCCO.CCCCCCCCCCCCCO.CCCCCCCCCCCCCCO
InChI
1S/C14H30O.C13H28O.C12H26O.C2H4O3.C2H6O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15;1-2-3-4-5-6-7-8-9-10-11-12-13-14;1-2-3-4-5-6-7-8-9-10-11-12-13;3-1-2(4)5;3-1-2-4/h15H,2-14H2,1H3;14H,2-13H2,1H3;13H,2-12H2,1H3;3H,1H2,(H,4,5);3-4H,1-2H2
InChI key
KQSLVJFTAGAPJL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service