Skip to Content
Merck
All Photos(1)

Key Documents

449725

Sigma-Aldrich

Cerium(III) bromide

anhydrous, powder, 99.9% trace metals basis

Synonym(s):

Cerium tribromide, Cerous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeBr3
CAS Number:
Molecular Weight:
379.83
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤2,000.0 ppm Trace Rare Earth Analysis

mp

730 °C (lit.)
730-732 °C (lit.)

SMILES string

Br[Ce](Br)Br

InChI

1S/3BrH.Ce/h3*1H;/q;;;+3/p-3

InChI key

MOOUSOJAOQPDEH-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service