Skip to Content
Merck
All Photos(2)

Key Documents

449253

Sigma-Aldrich

Ruthenium(III) iodide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RuI3 · H2O
CAS Number:
Molecular Weight:
499.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

reaction suitability

reagent type: catalyst
core: ruthenium

mp

590 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

O.I[Ru](I)I

InChI

1S/3HI.H2O.Ru/h3*1H;1H2;/q;;;;+3/p-3

InChI key

USLIQGJRXPCWLY-UHFFFAOYSA-K

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service