Skip to Content
Merck
All Photos(1)

Documents

448427

Sigma-Aldrich

(4R)-2-Hydroxy-5,5-dimethyl-4-phenyl-1,3,2-dioxaphosphorinan 2-oxide

98%

Synonym(s):

(2R,4R)-5,5-Dimethyl-2-hydroxy-4-phenyl-1,3,2-dioxaphosphorinan 2-oxide, (R)-(−)-2-Hydroxy-5,5-dimethyl-4-phenyl-1,3,2-dioxaphosphorinane 2-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15O4P
CAS Number:
Molecular Weight:
242.21
Beilstein:
4294714
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −62°, c = 0.5 in methanol

impurities

<5% water

mp

224-227 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)COP(O)(=O)O[C@@H]1c2ccccc2

InChI

1S/C11H15O4P/c1-11(2)8-14-16(12,13)15-10(11)9-6-4-3-5-7-9/h3-7,10H,8H2,1-2H3,(H,12,13)/t10-/m1/s1

InChI key

PSZSDCWNBXVDFG-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(4R)-2-Hydroxy-5,5-dimethyl-4-phenyl-1,3,2-dioxaphosphorinan 2-oxide can be used:
  • As a reactant in the phosphoalkoxylation of aromatic enones.
  • As a chiral resolving agent in the resolution of amino acids, amines, and amino alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiomeric impurities in chiral catalysts, auxiliaries and synthons used in enantioselective synthesis
Armstrong DW, et al.
Tetrahedron Asymmetry, 9(12), 2043-2064 (1998)
Four-component α-bromo-β-phosphoalkoxylation of aromatic α, β-unsaturated carbonyl compounds
Sohail M, et al.
Royal Society of Chemistry Advances, 5(22), 17014-17017 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service