Skip to Content
Merck
All Photos(1)

Key Documents

446351

Sigma-Aldrich

(S)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine

97%

Synonym(s):

(S)-1-Methyl-2-(piperidinomethyl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2
CAS Number:
Molecular Weight:
182.31
Beilstein:
4242955
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D −69°, c = 0.5 in ethanol

refractive index

n20/D 1.478 (lit.)

bp

121-123 °C/22 mmHg (lit.)

density

0.909 g/mL at 25 °C (lit.)

SMILES string

CN1CCC[C@H]1CN2CCCCC2

InChI

1S/C11H22N2/c1-12-7-5-6-11(12)10-13-8-3-2-4-9-13/h11H,2-10H2,1H3/t11-/m0/s1

InChI key

VCVPSRADUBPOKJ-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful chiral ligand for catalytic asymmetric aldol reactions. Mukaiyama, T.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 29, 59-59 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service