Skip to Content
Merck
All Photos(1)

Documents

442259

Sigma-Aldrich

Triisodecyl phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H21O)3P
CAS Number:
Molecular Weight:
502.79
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.46 (lit.)

bp

166 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCCCOP(OCCCCCCCC(C)C)OCCCCCCCC(C)C

InChI

1S/C30H63O3P/c1-28(2)22-16-10-7-13-19-25-31-34(32-26-20-14-8-11-17-23-29(3)4)33-27-21-15-9-12-18-24-30(5)6/h28-30H,7-27H2,1-6H3

InChI key

QEDNBHNWMHJNAB-UHFFFAOYSA-N

Application

Heat stabilizer for polymers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service