441120
3,3,5-Trimethylcyclohexyl methacrylate, mixture of isomers
98%
Synonym(s):
3,3,5-Trimethylcyclohexyl methacrylate, Isophoryl methacrylate
About This Item
Recommended Products
Assay
98%
form
liquid
contains
200 ppm monomethyl ether hydroquinone as inhibitor
refractive index
n20/D 1.456 (lit.)
bp
79 °C/1 mmHg (lit.)
mp
−15 °C (lit.)
density
0.93 g/mL at 25 °C (lit.)
SMILES string
CC1CC(CC(C)(C)C1)OC(=O)C(C)=C
InChI
1S/C13H22O2/c1-9(2)12(14)15-11-6-10(3)7-13(4,5)8-11/h10-11H,1,6-8H2,2-5H3
InChI key
DABQKEQFLJIRHU-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
208.4 °F - closed cup
Flash Point(C)
98 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service