Skip to Content
Merck
All Photos(1)

Key Documents

440280

Sigma-Aldrich

Ethyltrichlorosilane

99%

Synonym(s):

Dow Corning® product Z-1212, Trichloro(ethyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5Cl3Si
CAS Number:
Molecular Weight:
163.51
Beilstein:
1361384
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

99 °C (lit.)

mp

−106 °C (lit.)

density

1.238 g/mL at 25 °C (lit.)

SMILES string

CC[Si](Cl)(Cl)Cl

InChI

1S/C2H5Cl3Si/c1-2-6(3,4)5/h2H2,1H3

InChI key

ZOYFEXPFPVDYIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Repackaged by Aldrich Chemical Company, Inc. Manufactured by Dow Corning Corporation, U.S.A.
®Registered trademark of Dow Corning Corporation, U.S.A.
Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service