Skip to Content
Merck
All Photos(1)

Key Documents

437107

Sigma-Aldrich

Poly(vinylidene chloride-co-vinyl chloride)

powder, 240-320 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CCl2)x(CH2CHCl)y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

powder

composition

vinylidene chloride, ~65 wt. %

particle size

240-320 μm

melt viscosity

9,100 poise(185 °C) (100 sec-1)(lit.)

transition temp

Tm 164 °C

density

1.73 g/mL at 25 °C

SMILES string

ClC=C.ClC(Cl)=C

InChI

1S/C2H2Cl2.C2H3Cl/c1-2(3)4;1-2-3/h1H2;2H,1H2

InChI key

DHZSIQDUYCWNSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Superior gas and moisture barrier properties, self clinging.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 Oral

Target Organs

Liver, Nose

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Litauszki et al.
Journal of biomedical materials research, 35(1), 1-8 (1997-04-01)
Poly(ethylene oxide) (PEO) was immobilized by the Williamson ether synthesis onto halogenated surfaces such as poly(vinylidene chloride), surface-brominated polypropylene, and surface-brominated poly(ethylene terephthalate). PEO (Mw 20,000) was converted to its sodium salt and reacted for various times with the halogen-containing
Stephan Schreml et al.
Experimental dermatology, 20(7), 550-554 (2011-03-30)
In cutaneous wound healing, the role of oxygen in vivo is poorly understood. We studied wound surface pO(2) during physiological wound healing in humans. Split-thickness skin graft donor sites (n=12) served as standardized wound models. Wound surface pO(2) was measured
Nirav K Patel et al.
British journal of nursing (Mark Allen Publishing), 16(18), 1140-1142 (2007-12-13)
Clubfoot or congenital talipes equinovarus is a common condition affecting babies and non-surgical treatment involves serial manipulation and plaster casting for many weeks. The casts are susceptible to soiling during this time, which makes management and child care even more
S Baumgart
Pediatrics, 74(6), 1022-1028 (1984-12-01)
Eight very low-birth-weight premature infants (mean birth weight 1.11 +/- 0.05 [SEM]kg, mean gestation 30 +/- 1 weeks, and mean age 9 +/- 2 days) were studied under servocontrolled radiant warmers with and without a loosely fitted, transparent, and flexible
R C Strohman et al.
In vitro cellular & developmental biology : journal of the Tissue Culture Association, 26(2), 201-208 (1990-02-01)
Primary muscle cell cultures consisting of single myocytes and fibroblasts are grown on flexible, optically clear biomembranes. Muscle cell growth, fusion and terminal differentiation are normal. A most effective membrane for these cultures is commercially available Saran Wrap. Muscle cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service