Skip to Content
Merck
All Photos(2)

Key Documents

432962

Sigma-Aldrich

Vanadyl 2,11,20,29-tetra-tert-butyl-2,3-naphthalocyanine

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H56N8OV
CAS Number:
Molecular Weight:
1004.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 95%

mp

>300 °C (lit.)

λmax

808 nm

SMILES string

CC(C)(C)c1ccc2cc3c4nc(nc5n6[V](=O)n7c(nc8nc(nc6c9cc%10cc(ccc%10cc59)C(C)(C)C)c%11cc%12ccc(cc%12cc8%11)C(C)(C)C)c%13cc%14ccc(cc%14cc%13c7n4)C(C)(C)C)c3cc2c1

InChI

1S/C64H56N8.O.V/c1-61(2,3)41-17-13-33-25-45-49(29-37(33)21-41)57-65-53(45)70-58-51-31-39-23-43(63(7,8)9)19-15-35(39)27-47(51)55(67-58)72-60-52-32-40-24-44(64(10,11)12)20-16-36(40)28-48(52)56(68-60)71-59-50-30-38-22-42(62(4,5)6)18-14-34(38)26-46(50)54(66-59)69-57;;/h13-32H,1-12H3;;/q-2;;+2

InChI key

KYDACBKVLZCERN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service