Skip to Content
Merck
All Photos(2)

Documents

432210

Sigma-Aldrich

Zinc 2,11,20,29-tetra-tert-butyl-2,3-naphthalocyanine

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H56N8Zn
CAS Number:
Molecular Weight:
1002.57
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 90%

mp

>300 °C (lit.)

λmax

769 nm

SMILES string

CC(C)(C)c1ccc2cc3c4nc(nc5n6[Zn]n7c(nc8nc(nc6c9cc%10cc(ccc%10cc59)C(C)(C)C)c%11cc%12ccc(cc%12cc8%11)C(C)(C)C)c%13cc%14ccc(cc%14cc%13c7n4)C(C)(C)C)c3cc2c1

InChI

1S/C64H56N8.Zn/c1-61(2,3)41-17-13-33-25-45-49(29-37(33)21-41)57-65-53(45)70-58-51-31-39-23-43(63(7,8)9)19-15-35(39)27-47(51)55(67-58)72-60-52-32-40-24-44(64(10,11)12)20-16-36(40)28-48(52)56(68-60)71-59-50-30-38-22-42(62(4,5)6)18-14-34(38)26-46(50)54(66-59)69-57;/h13-32H,1-12H3;/q-2;+2

InChI key

QSAQNAXTLSXDAT-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yumiao Zhang et al.
Nature communications, 7, 11649-11649 (2016-05-20)
Injectable hydrophobic drugs are typically dissolved in surfactants and non-aqueous solvents which can induce negative side-effects. Alternatives like 'top-down' fine milling of excipient-free injectable drug suspensions are not yet clinically viable and 'bottom-up' self-assembled delivery systems usually substitute one solubilizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service